Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fk 453 (compound 1d) | 121524-18-3 | 99.2% | 375.46 g·mol⁻¹ | C23H25N3O2 | 5 MG
SDP

Catalog No. 5000244310
Encompass_Preferred

FK 453 (compound 1d) is a potent and selective adenosine A1 receptor antagonist (IC50 17.2 nM) intended for research use. It is a non-xanthine small molecule used in pharmacological studies to probe A1 receptor function and ligand interactions.

  • Potent adenosine A1 antagonist (IC50 17.2 nM).
  • High purity 99.2%.
  • Molecular weight 375.46 g·mol⁻¹.
  • Chemical formula C23H25N3O2.
  • Supplied as 5 mg solid for laboratory research.
  • Suitable for in vitro pharmacology and receptor binding assays.

Catalog No. 50-002-44310 Supplier Medchemexpress LLC Supplier No. HY105275A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FK 453 (compound 1d) is a potent and selective adenosine A1 receptor antagonist (IC50 17.2 nM) intended for research use. It is a non-xanthine small molecule used in pharmacological studies to probe A1 receptor function and ligand interactions.

  • Potent adenosine A1 antagonist (IC50 17.2 nM).
  • High purity 99.2%.
  • Molecular weight 375.46 g·mol⁻¹.
  • Chemical formula C23H25N3O2.
  • Supplied as 5 mg solid for laboratory research.
  • Suitable for in vitro pharmacology and receptor binding assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.