Learn More
AdipoGen FK-506,Chemical. CAS: 109581-93-3
SDP
Catalog No. 50464386
Encompass_Preferred
FK-506, AdipoGen Life Sciences. Chemical. CAS: 109581-93-3. Formula: C44H69NO12 . H2O. MW: 804.0 . 18.0. Isolated from Streptomyces FCZ0311. Potent immunosuppressant (as cyclosporin A and rapamycin). Suppresses proliferation of cytotoxic T cells and inhibits the production of T cell-derived mediators such as interleukin-2 (IL-2). Forms a complex with FK-506 binding protein 12 (FKBP12). Inhibits the activity of the calcium/calmodulin-dependent protein phosphatase 2B (PP2B; calcineurin), leading to disruption of T cell activation. Prevents rejection of transplanted organs. Anti-inflammatory compound in the treatment of several inflammatory skin diseases (e.g. atopic dermatitis) and with potential anti-rheumatic activity (rheumatoid arthritis). Neuroprotective. Regulates nitric oxide neurotoxicity. Neurotransmitter. Ca2+ release compound. NF-kappaB suppressor by induction of unfolded protein response (UPR). Anti-cancer compound. Apoptosis inducer.
Catalog No. 50-464-386 Supplier AdipoGen Supplier No. AGCN20047M025
FK-506, AdipoGen Life Sciences. Chemical. CAS: 109581-93-3. Formula: C44H69NO12 . H2O. MW: 804.0 . 18.0. Isolated from Streptomyces FCZ0311. Potent immunosuppressant (as cyclosporin A and rapamycin). Suppresses proliferation of cytotoxic T cells and inhibits the production of T cell-derived mediators such as interleukin-2 (IL-2). Forms a complex with FK-506 binding protein 12 (FKBP12). Inhibits the activity of the calcium/calmodulin-dependent protein phosphatase 2B (PP2B; calcineurin), leading to disruption of T cell activation. Prevents rejection of transplanted organs. Anti-inflammatory compound in the treatment of several inflammatory skin diseases (e.g. atopic dermatitis) and with potential anti-rheumatic activity (rheumatoid arthritis). Neuroprotective. Regulates nitric oxide neurotoxicity. Neurotransmitter. Ca2+ release compound. NF-kappaB suppressor by induction of unfolded protein response (UPR). Anti-cancer compound. Apoptosis inducer.