Learn More
Carbosynth LLC. FK-506 (Tacrolimus), 1000mg, CAS# 104987-11-3, MF= C44H69NO12
SDP
Supplier:  Carbosynth LLC. INT3098PI1000MG
Encompass_Preferred
Tacrolimus, Application: Induces Immunosuppression By Inhibiting T Cell Proliferation; Inhibits FKBP12 and Calcineurin (IC50 approx 1 nM); Cell Permeable, Notes: FK-506 is cell permeable and induces immunosuppression by inhibiting T cell proliferation. It also inhibits FKBP12 and calcineurin (IC50 approx 1 nM).Solubilization: May be dissolved in DMSO (50 mg/ml); or ethanol (50 mg/ml), salt: , origin/species: Produced by fermentation, MW: 804
Catalog No. 50-195-2410