Learn More
AdipoGen FK-866 (5 mg)
SDP
Supplier:  AdipoGen AGCR10011M005
Encompass_Preferred
Chemical. CAS: 658084-64-1. Formula: C24H29N3O2. MW: 391.5. Nampt/visfatin inhibitor. Inhibitor of NAD+ biosynthesis. Inhibits the enzyme/substrate complex and the free enzyme (Ki = 0.4 nM and Ki = 0.3 nM, respectively). Apoptosis inducer. Autophagy inducer. Causes premature senescence. Angiogenesis inhibitor.
Catalog No. 50-205-3760