Learn More
AdipoGen FK-866
SDP
Supplier:  AdipoGen AGCR10011M001
Encompass_Preferred
Chemical. CAS 658084-64-1. Formula C24H29N3O2. MW 391.5. Nampt/visfatin inhibitor. Inhibitor of NAD biosynthesis. Inhibits the enzyme/substrate complex and the free enzyme Ki = 0.4 nM and Ki = 0.3 nM, respectively. Apoptosis inducer. Autophagy inducer. Causes premature senescence. Angiogenesis inhibitor.
Catalog No. 50-256-2397