Learn More
Apexbio Technology LLC FK 866 hydrochloride(Synonyms: FK866 hydrochloride, APO866 hydrochloride, Daporinad hydrochloride), 10mM (in 1mL DMSO), CAS: 658084-64-1 (free base).
SDP
Catalog No. 5000060113
Encompass_Preferred
FK 866 hydrochloride (CAS 658084-64-1 free base) is a potent inhibitor of nicotinamide phosphoribosyltransferase (NMPRTase) an enzyme essential for NAD biosynthesis exhibiting activity with IC50 values ranging from 0 09 nM to 27 2 nM By blocking NAD synthesis FK 866 disrupts ATP production induces mitochondria membrane depolarization and triggers autophagy independent of caspase activation It selectively targets hematologic cancer cells such as AML lines sparing normal hematopoietic progenitor cells FK 866 demonstrates antitumor efficacy in vivo significantly suppressing growth in AML-M4 and Namalwa xenograft models
Catalog No. 50-000-60113 Supplier Apexbio Technology LLC Supplier No. A86315.1
FK 866 hydrochloride (CAS 658084-64-1 free base) is a potent inhibitor of nicotinamide phosphoribosyltransferase (NMPRTase) an enzyme essential for NAD biosynthesis exhibiting activity with IC50 values ranging from 0 09 nM to 27 2 nM By blocking NAD synthesis FK 866 disrupts ATP production induces mitochondria membrane depolarization and triggers autophagy independent of caspase activation It selectively targets hematologic cancer cells such as AML lines sparing normal hematopoietic progenitor cells FK 866 demonstrates antitumor efficacy in vivo significantly suppressing growth in AML-M4 and Namalwa xenograft models