Learn More
STEMCELL Technologies FK506, Size: 10 mg
SDP
Catalog No. 501976189
Encompass
FK506 is a macrolide antibiotic with potent immunosuppressive properties (Thomson et al.). It inhibits the calcineurin/NFAT pathway (Martinez-Martinez & Redondo) and binds with high affinity to FK506 binding protein 12 (FKBP-12) (Thomson et al.).MAINTENANCE AND SELF-RENEWAL· Promotes the self-renewal of mouse embryonic stem cells independent of leukemia inhibitory factor (LIF), and attenuates differentiation following LIF withdrawal (Li et al.). · In combination with 1-Azakenpaullone (Catalog #74252) and ID-8 (Catalog #72502), enhances the expansion rates of several human pluripotent stem cell lines (Yasuda et al.).IMMUNOLOGY· Inhibits pro-inflammatory cytokine production by activated T cells (Sakuma et al.).
Catalog No. 50-197-6189 Supplier STEMCELL Technologies Supplier No. 74152
FK506 is a macrolide antibiotic with potent immunosuppressive properties (Thomson et al.). It inhibits the calcineurin/NFAT pathway (Martinez-Martinez & Redondo) and binds with high affinity to FK506 binding protein 12 (FKBP-12) (Thomson et al.).MAINTENANCE AND SELF-RENEWAL· Promotes the self-renewal of mouse embryonic stem cells independent of leukemia inhibitory factor (LIF), and attenuates differentiation following LIF withdrawal (Li et al.). · In combination with 1-Azakenpaullone (Catalog #74252) and ID-8 (Catalog #72502), enhances the expansion rates of several human pluripotent stem cell lines (Yasuda et al.).IMMUNOLOGY· Inhibits pro-inflammatory cytokine production by activated T cells (Sakuma et al.).