Learn More
Abcam FK506 (Tacrolimus), Immunosuppressant, 10MG
Catalog No. 502584648
Encompass_Preferred
MW 804 Da, Purity >98%. Immunosuppressant. Complexes with FKBP-12 to inhibit calcineurin. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4648 Supplier Abcam Supplier No. AB12022310MG
MW 804 Da, Purity >98%. Immunosuppressant. Complexes with FKBP-12 to inhibit calcineurin. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement