Learn More
Abcam FK506 (Tacrolimus), Immunosuppressant, 50MG
Catalog No. 502584647
Encompass_Preferred
MW 804 Da, Purity >98%. Immunosuppressant. Complexes with FKBP-12 to inhibit calcineurin. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4647 Supplier Abcam Supplier No. AB12022350MG
MW 804 Da, Purity >98%. Immunosuppressant. Complexes with FKBP-12 to inhibit calcineurin. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement