Learn More
Medchemexpress LLC HY-50876 100mg , FK866 Daporinad;APO866 CAS:658084-64-1 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY50876100MG
Encompass_Preferred
Medchemexpress, HY-50876 100mg FK866 Daporinad;APO866 CAS:658084-64-1 Formula:C24H29N3O2 IC50: 0.09 nM (NMPRTase) Purity:98% FK866 is an effective inhibitor of nicotinamide phosphoribosyltransferase ( NMPRTase ) with IC 50 of 0.09 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2806