Learn More
Aviva Systems Biology Corp FKBP10 Antibody - C-terminal region (ARP65075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP10. It was validated on Western Blot. . Alternate names: FKBP65, FLJ20683, FLJ22041, FLJ23833, PPIASE, hFKBP65, OI6, OI11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-421