Learn More
Medchemexpress LLC HY-114421 5mg , FKBP12 PROTAC dTAG-13 CAS:2064175-41-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1144215MG
Encompass_Preferred
HY-114421 5mg FKBP12 PROTAC dTAG-13 CAS: 2064175-41-1 FKBP12 PROTAC dTAG-13 (dTAG-13), a PROTAC-based heterobifunctional degrader, is a selective degrader of FKBP12F36V with expression of FKBP12F36V in-frame with a protein of interest. FKBP12 PROTAC dTAG-13 effectively engages FKBP12F36V and CRBN, thereby selectively degrading FKBP12F36V. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1677