Learn More
Medchemexpress LLC FKBP12 PROTAC RC32 | 2375555-66-9 | 97.0% | 100 MG
SDP
Catalog No. 5000339946
Encompass_Preferred
FKBP12 PROTAC RC32 (RC32) is a potent FKBP12 degrader that utilizes PROTAC technology. It is a conjugate of Rapamycin and a ligand for a Cereblon E3 ubiquitin ligase, designed to efficiently degrade FKBP12 protein.
  • Potent degrader of FKBP12
  • Based on PROTAC technology
  • Contains conjugation of Rapamycin and a ligand for a Cereblon E3 ubiquitin ligase
  • Efficiently degrades FKBP12 protein in most organs (in vivo studies)
  • Induces FKBP12 protein degradation in human Jurkat cells (DC50 ~0.3 nM)
Catalog No. 50-003-39946 Supplier Medchemexpress LLC Supplier No. HY130835100MG
FKBP12 PROTAC RC32 (RC32) is a potent FKBP12 degrader that utilizes PROTAC technology. It is a conjugate of Rapamycin and a ligand for a Cereblon E3 ubiquitin ligase, designed to efficiently degrade FKBP12 protein.
  • Potent degrader of FKBP12
  • Based on PROTAC technology
  • Contains conjugation of Rapamycin and a ligand for a Cereblon E3 ubiquitin ligase
  • Efficiently degrades FKBP12 protein in most organs (in vivo studies)
  • Induces FKBP12 protein degradation in human Jurkat cells (DC50 ~0.3 nM)