Learn More
Medchemexpress LLC FKBP12 PROTAC RC32 | 2375555-66-9 | 98.6% | C75H107N7O20 | 10MG
SDP
Catalog No. 5000260174
Encompass_Preferred
FKBP12 PROTAC RC32 is a proteolysis targeting chimera (PROTAC) designed to induce selective ubiquitin-proteasome degradation of FKBP12 for target validation and mechanism-of-action studies in cellular and in vivo models. It links an FKBP12-binding ligand to an E3 ligase recruiting moiety to promote efficient target knockdown.
  • Potent and selective FKBP12 degradation in cells.
  • Enables in vivo target knockdown studies.
  • Suitable for target validation and mechanism studies.
  • Supplied as a powder with long-term storage recommendations.
  • Soluble in DMSO at 10 mg/mL; ultrasonic assistance recommended.
  • High reported purity suitable for biological assays.
Catalog No. 50-002-60174 Supplier Medchemexpress LLC Supplier No. HY13083510MG
FKBP12 PROTAC RC32 is a proteolysis targeting chimera (PROTAC) designed to induce selective ubiquitin-proteasome degradation of FKBP12 for target validation and mechanism-of-action studies in cellular and in vivo models. It links an FKBP12-binding ligand to an E3 ligase recruiting moiety to promote efficient target knockdown.
  • Potent and selective FKBP12 degradation in cells.
  • Enables in vivo target knockdown studies.
  • Suitable for target validation and mechanism studies.
  • Supplied as a powder with long-term storage recommendations.
  • Soluble in DMSO at 10 mg/mL; ultrasonic assistance recommended.
  • High reported purity suitable for biological assays.