Learn More
Medchemexpress LLC HY-130835 5mg Medchemexpress, FKBP12 PROTAC RC32 CAS:2375555-66-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1308355MG
Encompass_Preferred
Medchemexpress, HY-130835 5mg FKBP12 PROTAC RC32 CAS:2375555-66-9 FKBP12 PROTAC RC32 (RC32) is a potent FKBP12 degrader based on PROTAC technology. FKBP12 PROTAC RC32 contains conjugation of Rapamycin (HY-10219) and a ligand for an E3 ubiquitin ligase (Pomalidomide; HY-10984). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1254