Learn More
Aviva Systems Biology Corp FKBP14 Antibody - middle region (ARP62658_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP14. It was validated on Western Blot. . Alternate names: FKBP22, FLJ20731ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-732