Learn More
Sino Biological FKBP14 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 13522T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FKBP14 (rh FKBP14; Catalog#13522-H08H; Q9NWM8; Met1-Lys207). FKBP14 specific IgG was purified by Human FKBP14 affinity chromatography.
Catalog No. 50-224-6393