Learn More
Aviva Systems Biology Corp FKBP1A Antibody - N-terminal region (ARP65168_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65168_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP1A. It was validated on Western Blot. . Alternate names: FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE, FKBP-1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-447