Learn More
Aviva Systems Biology Corp FKBP4 Antibody - C-terminal region (ARP30180_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30180_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FKBP4. Alternate names: FKBP52, FKBP59, HBI, Hsp56, PPIase, p52, FKBP51ApplicationWB, IHC. Species reactivity: Human, Pig
Catalog No. 50-014-54140