Learn More
ACROBiosystems ACROBiosystems Human FKBP4 / FKBP52 Protein, His,PA Tag
Catalog No. 501957308
Encompass_Preferred
Recombinant Protein;50UG;Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 459 (Accession # AAH01786).;This protein carries a polyhistidine tag at the C-terminus, followed by a PA tag, and has a calculated MW of 55.1 kDa. The reducing (R) protein migrates as 60 kDa in SDS-PAGE.
Catalog No. 50-195-7308 Supplier ACROBiosystems Supplier No. FK4H52Q350UG
Recombinant Protein;50UG;Human FKBP4, His,PA Tag (FK4-H52Q3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 459 (Accession # AAH01786).;This protein carries a polyhistidine tag at the C-terminus, followed by a PA tag, and has a calculated MW of 55.1 kDa. The reducing (R) protein migrates as 60 kDa in SDS-PAGE.