Learn More
Enzo Life Sciences FKBP59 monoclonal antibody (KN382/EC1) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISRA1400F
Encompass_Preferred
Clone: KN382/EC1. Host: Mouse. Isotype: IgG1. Immunogen: Partially purified 9S rabbit uterine progesterone receptor. UniProt ID: P27124. Source: Purified from mouse ascites. Species reactivity: Human, Mouse Rabbit, Monkey, Mink, Hamster, Guinea pig, Bovine. Applications: IHC, IP, WB. Recommended Dilutions: Immunoprecipitation (10µg/ml) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7672