Learn More
Aviva Systems Biology Corp FKBP6 Antibody - middle region (ARP43027_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43027_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP6. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human FKBP6. Alternate names: FKBP36, MGC87179, PPIaseApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58399