Learn More
Aviva Systems Biology Corp FKBP8 Antibody - N-terminal region (ARP46862_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46862_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBP8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FKBP8. Alternate names: FKBP38, FKBPr38ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59423