Learn More
Aviva Systems Biology Corp FKBPL Antibody - N-terminal region (ARP57597_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57597_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FKBPL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FKBPL. Alternate names: DIR1, NG7, WISP39ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-63025