Learn More
Aviva Systems Biology Corp Fkrp Antibody - C-terminal region (ARP44898_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44898_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Fkrp. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: A830029B19Rik, AI842067, AI847300, LGMD1I, MDC1CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-166