Learn More
Medchemexpress LLC HY-111102 5mg Medchemexpress, FL-411 CAS:2118944-88-8 Purity:>98%
SDP
Catalog No. 501968821
Encompass_Preferred
Medchemexpress, HY-111102 5mg FL-411 CAS:2118944-88-8 FL-411 is a potent and selective BRD4 inhibitor with an IC50 of 0.43±0.09 μM for BRD4(1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8821 Supplier Medchemexpress LLC Supplier No. HY1111025MG
Medchemexpress, HY-111102 5mg FL-411 CAS:2118944-88-8 FL-411 is a potent and selective BRD4 inhibitor with an IC50 of 0.43±0.09 μM for BRD4(1). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.