Learn More
Sorbent Technologies Inc Purity Flash Cartridges, C18 Spherical Silica Gel, Premium Rf, 100A, 20-45um, 33g, 1/pk
SDP
Catalog No. 501139152
Encompass
Purity™ Cartridges are superior because they're packed with Premium Rf™ C18 Spherical Silica Gel. The porosity is 100A, particle distribution is 20-45um, bed mass is 33g, and quantity is 1 per pack. Our packing technique and Premium Rf's narrow particle distribution results in a uniform packed bed. The benefits are no channeling, higher resolution, decrease in backpressure, faster flow, and faster run time. Additionally, the small pore volume allows for more silica gel per column volume leading to greater loadability and less solvent use. Finally, the high purity silica gel (minimal surface metals) reduces mixed-mode interactions (no peak tailing or resolution loss). The polypropylene cartridge has standard luer-lock end fittings and is spin-welded (leak-free) allowing higher pressures up to 400 psi. The cartridges are compatible with all flash chromatography systems.
Catalog No. 50-113-9152 Supplier Sorbent Technologies Inc Supplier No. CFC736460251
Purity™ Cartridges are superior because they're packed with Premium Rf™ C18 Spherical Silica Gel. The porosity is 100A, particle distribution is 20-45um, bed mass is 33g, and quantity is 1 per pack. Our packing technique and Premium Rf's narrow particle distribution results in a uniform packed bed. The benefits are no channeling, higher resolution, decrease in backpressure, faster flow, and faster run time. Additionally, the small pore volume allows for more silica gel per column volume leading to greater loadability and less solvent use. Finally, the high purity silica gel (minimal surface metals) reduces mixed-mode interactions (no peak tailing or resolution loss). The polypropylene cartridge has standard luer-lock end fittings and is spin-welded (leak-free) allowing higher pressures up to 400 psi. The cartridges are compatible with all flash chromatography systems.