Learn More
Molecular Devices Molecular Devices Pipettor Head Kit, 8-Channel, 96-Well, Tips Included
Catalog No. 5000014542
Encompass_Preferred
The Molecular Devices FlexStation 3 Pipettor Head Kit supplies an integrated 8-channel pipettor and starter consumables to enable automated liquid handling on FlexStation 3 multi-mode microplate readers. It is intended for 96-well assay workflows, supporting compound additions and real-time kinetic measurements.
  • 8-channel pipettor
  • Ten racks of 96-well pipet tips, black
  • One 96-well yellow plate
  • Designed for automated liquid handling of 96-well assays
  • Supports real-time kinetic compound additions
Catalog No. 50-000-14542 Supplier Molecular Devices Supplier No. 02006182
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices FlexStation 3 Pipettor Head Kit supplies an integrated 8-channel pipettor and starter consumables to enable automated liquid handling on FlexStation 3 multi-mode microplate readers. It is intended for 96-well assay workflows, supporting compound additions and real-time kinetic measurements.
  • 8-channel pipettor
  • Ten racks of 96-well pipet tips, black
  • One 96-well yellow plate
  • Designed for automated liquid handling of 96-well assays
  • Supports real-time kinetic compound additions