Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fl118-14-propanol | 2821768-98-1 | 98.7% | 450.44 | C24H22N2O7 | 10 MG
SDP

Catalog No. 5000204338
Encompass_Preferred

FL118-14-propanol is a synthetic derivative of FL118 developed as an antibody-drug conjugate (ADC) payload. It has demonstrated antitumor activity in mice and is supplied for research use only.

  • CAS number 2821768-98-1.
  • Molecular formula C24H22N2O7.
  • Molecular weight 450.44 g/mol.
  • Purity 98.7%.
  • Appearance solid, light yellow to yellow.
  • Soluble in DMSO (100 mg/mL); hygroscopic DMSO may affect solubility.
  • Storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-04338 Supplier Medchemexpress LLC Supplier No. HY15685610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FL118-14-propanol is a synthetic derivative of FL118 developed as an antibody-drug conjugate (ADC) payload. It has demonstrated antitumor activity in mice and is supplied for research use only.

  • CAS number 2821768-98-1.
  • Molecular formula C24H22N2O7.
  • Molecular weight 450.44 g/mol.
  • Purity 98.7%.
  • Appearance solid, light yellow to yellow.
  • Soluble in DMSO (100 mg/mL); hygroscopic DMSO may affect solubility.
  • Storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.