Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fl118-14-propanol | 2821768-98-1 | 98.7% | 450.44 g/mol | C24H22N2O7 | 25 MG
SDP

Catalog No. 5000204397
Encompass_Preferred

FL118-14-Propanol is a derivative of FL118 described as an ADC molecule with reported anti-tumor activity in mice. It is supplied as a solid in multiple milligram pack sizes and as a prepared DMSO solution for research use.

  • CAS number 2821768-98-1.
  • Purity 98.7%.
  • Molecular weight 450.44 g/mol.
  • Chemical formula C24H22N2O7.
  • Available as solid (5, 10, 25, 50, 100 mg) and as 10 mM in DMSO solution.
  • Soluble in DMSO.
  • Reported anti-tumor effect in mice.

Catalog No. 50-002-04397 Supplier Medchemexpress LLC Supplier No. HY15685625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FL118-14-Propanol is a derivative of FL118 described as an ADC molecule with reported anti-tumor activity in mice. It is supplied as a solid in multiple milligram pack sizes and as a prepared DMSO solution for research use.

  • CAS number 2821768-98-1.
  • Purity 98.7%.
  • Molecular weight 450.44 g/mol.
  • Chemical formula C24H22N2O7.
  • Available as solid (5, 10, 25, 50, 100 mg) and as 10 mM in DMSO solution.
  • Soluble in DMSO.
  • Reported anti-tumor effect in mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.