Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fl118-14-propanol | 2821768-98-1 | 98.7% | 450.44 g/mol | C24H22N2O7 | 50 MG
SDP

Catalog No. 5000204478
Encompass_Preferred

FL118-14-propanol is a synthetic derivative of FL118 developed as an ADC payload candidate with reported anti-tumor activity in mice; supplied for laboratory research use only.

  • Provides an ADC payload scaffold for preclinical anti-tumor research.
  • High purity: 98.7%.
  • Formula: C24H22N2O7; molecular weight: 450.44 g/mol.
  • CAS number: 2821768-98-1.
  • Available in multiple sizes, including 50 MG solid packs.
  • Intended for research use only; not for clinical or diagnostic use.

Catalog No. 50-002-04478 Supplier Medchemexpress LLC Supplier No. HY15685650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FL118-14-propanol is a synthetic derivative of FL118 developed as an ADC payload candidate with reported anti-tumor activity in mice; supplied for laboratory research use only.

  • Provides an ADC payload scaffold for preclinical anti-tumor research.
  • High purity: 98.7%.
  • Formula: C24H22N2O7; molecular weight: 450.44 g/mol.
  • CAS number: 2821768-98-1.
  • Available in multiple sizes, including 50 MG solid packs.
  • Intended for research use only; not for clinical or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.