Learn More
Medchemexpress LLC Fl118-14-propanol-d5 | 2821768-98-1 | 95.4% | 455.47 | C24H17D5N2O7 | 10 MG
SDP
Catalog No. 5000287735
Encompass_Preferred
FL118-14-Propanol-d5 is a deuterium-labeled research compound used as a stable-isotope tracer and internal standard for quantitative analytical methods and preclinical studies. It is intended for research use only and supports pharmacokinetic, metabolic profiling, and ADC-related investigations. The material is supplied as a solid with specified purity and defined storage recommendations for stability.
  • Deuterium-labeled internal standard for quantitative NMR, LC-MS, and GC-MS assays.
  • Intended for research use only; not for human or clinical use.
  • Useful for pharmacokinetic and metabolic profiling studies.
  • Applicable to ADC synthesis and preclinical antitumor research.
  • Supplied with reported purity of approximately 95.4%.
  • Store sealed at -20°C; in solvent, store at -80°C for up to 6 months or -20°C for 1 month.
  • Available in multiple pack sizes, including 10 mg, to support small-scale analytical work.
Catalog No. 50-002-87735 Supplier Medchemexpress LLC Supplier No. HY156856S10MG
FL118-14-Propanol-d5 is a deuterium-labeled research compound used as a stable-isotope tracer and internal standard for quantitative analytical methods and preclinical studies. It is intended for research use only and supports pharmacokinetic, metabolic profiling, and ADC-related investigations. The material is supplied as a solid with specified purity and defined storage recommendations for stability.
  • Deuterium-labeled internal standard for quantitative NMR, LC-MS, and GC-MS assays.
  • Intended for research use only; not for human or clinical use.
  • Useful for pharmacokinetic and metabolic profiling studies.
  • Applicable to ADC synthesis and preclinical antitumor research.
  • Supplied with reported purity of approximately 95.4%.
  • Store sealed at -20°C; in solvent, store at -80°C for up to 6 months or -20°C for 1 month.
  • Available in multiple pack sizes, including 10 mg, to support small-scale analytical work.