Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fl118-C3-O-C-amide-C-NH2-d5 formate | 2821769-41-7 | 587.59 g/mol | C28H25D5N4O10 | 10 MG
SDP

Catalog No. 5000288746
Encompass_Preferred

FL118-C3-O-C-amide-C-NH2-d5 formate is a deuterium-labeled formate salt of an FL118-derived ADC linker intended for research use as a tracer, internal standard, or building block in antibody-drug conjugate synthesis. It is supplied in small milligram quantities for analytical and synthetic workflows.

  • Deuterium-labeled (d5) tracer for quantitative analyses.
  • Formate salt for improved handling and solubility.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Available in milligram-scale quantities for research applications.
  • Molecular formula C28H25D5N4O10 and molecular weight 587.59 g/mol.

Catalog No. 50-002-88746 Supplier Medchemexpress LLC Supplier No. HY161022S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FL118-C3-O-C-amide-C-NH2-d5 formate is a deuterium-labeled formate salt of an FL118-derived ADC linker intended for research use as a tracer, internal standard, or building block in antibody-drug conjugate synthesis. It is supplied in small milligram quantities for analytical and synthetic workflows.

  • Deuterium-labeled (d5) tracer for quantitative analyses.
  • Formate salt for improved handling and solubility.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Available in milligram-scale quantities for research applications.
  • Molecular formula C28H25D5N4O10 and molecular weight 587.59 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.