Learn More
Medchemexpress LLC FL118-C3-O-C-amide-C-NH2-d5 formate | 587.59 | 5 MG
SDP
Catalog No. 5000395351
Encompass_Preferred
FL118-C3-O-C-amide-C-NH2-d5 formate is a deuterium-labeled FL118-C3-O-C-amide-C-NH2 formate, an ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Used as an ADC linker
  • Applicable in the synthesis of antibody-drug conjugates (ADCs)
  • Heavy isotopes can serve as tracers for quantitation during drug development
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-95351 Supplier Medchemexpress LLC Supplier No. HY161022S15MG
FL118-C3-O-C-amide-C-NH2-d5 formate is a deuterium-labeled FL118-C3-O-C-amide-C-NH2 formate, an ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Used as an ADC linker
  • Applicable in the synthesis of antibody-drug conjugates (ADCs)
  • Heavy isotopes can serve as tracers for quantitation during drug development
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs