Learn More
Julabo USA Inc Each, FL1201 chiller, -20+40C, 1.2 kW, 115V/20A/60Hz
SDP
Catalog No. 507533948
Encompass_Preferred
Chillers, Julabo, FL1201 Recirculating Cooler -20…+40 °C., 1.2 kW cooling power; ±0.5°C stability. Proportional cooling control adjusts the cooling power to the load of the application saving energy. Units incorporate RS232 interface and alarm output for PC integration and control. Models include a splash-proof membrane keypad with LED temperature display. Easy filling / draining with a fill level indicator and overflow port on the back. Units constructed of stainless steel and high-grade plastic with stainless steel internal tank. Centrifugal pressure pump. Units with 3 and 6 bar pump include internal by-pass for pressure adjustment with analog pressure gaugeMay be used with water, water/glycol mixtures and silicone fluids.The FL300, FL601, FL1201, FL1701, FLW1701 come with 8 and 12 mm hose barbs. The FLxxx3 models come with 3/4″ hose barbs. The FLxxx6 models come with 1″ hose barbs. Included with each unit: 2 barbed fittings for tubing 8/12mm ID. (pump connections with M16x1 male)
Catalog No. 50-753-3948 Supplier Julabo USA Inc Supplier No. 9661012.2
Add to Cart
Add to Cart
Request a Quote
Chillers, Julabo, FL1201 Recirculating Cooler -20…+40 °C., 1.2 kW cooling power; ±0.5°C stability. Proportional cooling control adjusts the cooling power to the load of the application saving energy. Units incorporate RS232 interface and alarm output for PC integration and control. Models include a splash-proof membrane keypad with LED temperature display. Easy filling / draining with a fill level indicator and overflow port on the back. Units constructed of stainless steel and high-grade plastic with stainless steel internal tank. Centrifugal pressure pump. Units with 3 and 6 bar pump include internal by-pass for pressure adjustment with analog pressure gaugeMay be used with water, water/glycol mixtures and silicone fluids.The FL300, FL601, FL1201, FL1701, FLW1701 come with 8 and 12 mm hose barbs. The FLxxx3 models come with 3/4″ hose barbs. The FLxxx6 models come with 1″ hose barbs. Included with each unit: 2 barbed fittings for tubing 8/12mm ID. (pump connections with M16x1 male)