Learn More
Sino Biological Anti-flagellin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 40067T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Listeria monocytogenes flagellin / FlaA (Catalog#40067-V07E; YP_013330.1; Met1-Ser287). Listeria monocytogenes flagellin / FlaA specific IgG was purified by Listeria monocytogenes flagellin / FlaA affinity chromatography.
Catalog No. 50-255-3743