Learn More
CAPRICO BIOTECHNOLOGIES INC Flaer mFluor450-4105141
Catalog No. 502279299
Encompass_Preferred
The Flaer mFluor450-4105141 is a fluorescent dye from Caprico Bio that has an excitation maximum at 405 nm and emission maximum at 450 nm. This organic dye is compatible with flow cytometry and fluorescence microscopy applications. It has a molecular weight of 732.9 g/mol and is supplied as a lyophilized sodium salt that is soluble in water and PBS. When conjugated to antibodies or other biomolecules, mFluor450-4105141 enables fluorescent detection and analysis. This bright, photostable dye is offered in pack sizes of 1 mg, 5 mg, and 25 mg.
This item is not returnable. View return policy
Catalog No. 50-227-9299 Supplier CAPRICO BIOTECHNOLOGIES INC Supplier No. 4105141
The Flaer mFluor450-4105141 is a fluorescent dye from Caprico Bio that has an excitation maximum at 405 nm and emission maximum at 450 nm. This organic dye is compatible with flow cytometry and fluorescence microscopy applications. It has a molecular weight of 732.9 g/mol and is supplied as a lyophilized sodium salt that is soluble in water and PBS. When conjugated to antibodies or other biomolecules, mFluor450-4105141 enables fluorescent detection and analysis. This bright, photostable dye is offered in pack sizes of 1 mg, 5 mg, and 25 mg.