Learn More
Genscript Corporation DYKDDDDK Tag Antibody Plate (White, 96-well)
Catalog No. NC1503986
Encompass
DYKDDDDK (Flag) Tag Antibody Plate is a microtiter plate coated with THETM Anti DYKDDDDK (Flag) Tag Antibody. The plate is designed for the rapid capturing of DYKDDDDK (Flag) tagged protein expressed in different systems, including, E. coli, yeast and mammalian extracts and cell culture supernatants. It can be applied for the detection of DYKDDDDK (Flag) tag located at N-terminus, C-terminus, or internal of the protein. The plate demonstrates high sensitivity and high binding capacity. The capacity and sensitivity varies, depending on protein size, structure and solution buffer. Generally, proteins with lower molecular weight (M.W) can be more sensitive to the DYKDDDDK (Flag) antibody plate than ones with higher M.W. The plate is tolerable with several common used reagents. Avoid using detection reagents that can recognize mouse IgG, such as anti mouse IgG antibodies and protein A.
Catalog No. NC1503986 Supplier Genscript Corporation Supplier No. L00455W
DYKDDDDK (Flag) Tag Antibody Plate is a microtiter plate coated with THETM Anti DYKDDDDK (Flag) Tag Antibody. The plate is designed for the rapid capturing of DYKDDDDK (Flag) tagged protein expressed in different systems, including, E. coli, yeast and mammalian extracts and cell culture supernatants. It can be applied for the detection of DYKDDDDK (Flag) tag located at N-terminus, C-terminus, or internal of the protein. The plate demonstrates high sensitivity and high binding capacity. The capacity and sensitivity varies, depending on protein size, structure and solution buffer. Generally, proteins with lower molecular weight (M.W) can be more sensitive to the DYKDDDDK (Flag) antibody plate than ones with higher M.W. The plate is tolerable with several common used reagents. Avoid using detection reagents that can recognize mouse IgG, such as anti mouse IgG antibodies and protein A.