Learn More
Sino Biological Anti-DYKDDDDK (FLAG epitope Tag) Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 100233MM01H20
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, the synthetic peptide corresponding to the DYKDDDDK-tag sequence (CDYKDDDDK). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-1424