Learn More
Enzo Life Sciences FLAG polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADISAB4100100
Encompass_Preferred
Host: Rabbit. Isotype: IgG. Immunogen: Synthetic peptide (CDYKDDDDK) coupled to KLH. Source: Purified from rabbit serum. Species reactivity: Species independent. Specificity: Recognizes FLAG-tagged proteins. Applications: ELISA, WB. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-7396