Learn More
Medchemexpress LLC HY-P0223 25mg Medchemexpress, FLAG peptide CAS:98849-88-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP022325MG
Encompass_Preferred
Medchemexpress, HY-P0223 25mg FLAG peptide CAS:98849-88-8 FLAG peptide is an eight amino acids peptide (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) with an enterokinase-cleavage site; designed for antibody-mediated identification and purification of recombinant proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9431