Learn More
Medchemexpress LLC FLAG peptide TFA | 99.6% | 1012.97 | 5 MG
SDP
Catalog No. 5000426752
Encompass_Preferred
FLAG peptide TFA is the trifluoroacetic acid salt form of FLAG peptide, often denoted as HY-P0223. It functions as a versatile fusion tag specifically designed for the purification of recombinant proteins. This peptide plays a crucial role in maintaining the natural, functional folding of the proteins it is fused with, ensuring their biological activity. It can be efficiently removed by enterokinase and is capable of being eluted under non-denaturing conditions, which is beneficial for preserving protein integrity.
  • Multifunctional fusion tag
  • Used for purification of recombinant proteins
  • Maintains natural folding of fusing proteins
  • Can be removed by enterokinase
  • Eluted under non-denaturing conditions
Catalog No. 50-004-26752 Supplier Medchemexpress LLC Supplier No. HYP0223A5MG
FLAG peptide TFA is the trifluoroacetic acid salt form of FLAG peptide, often denoted as HY-P0223. It functions as a versatile fusion tag specifically designed for the purification of recombinant proteins. This peptide plays a crucial role in maintaining the natural, functional folding of the proteins it is fused with, ensuring their biological activity. It can be efficiently removed by enterokinase and is capable of being eluted under non-denaturing conditions, which is beneficial for preserving protein integrity.
  • Multifunctional fusion tag
  • Used for purification of recombinant proteins
  • Maintains natural folding of fusing proteins
  • Can be removed by enterokinase
  • Eluted under non-denaturing conditions