Learn More
Medchemexpress LLC FLAG peptide TFA | 99.6% | 1012.97 (free base) | 25 MG
SDP
Catalog No. 5000426469
Encompass_Preferred
FLAG peptide TFA is the TFA salt form of FLAG peptide. It is a multifunctional fusion tag used for the purification of recombinant proteins. It maintains the natural folding of its fusing proteins and can be removed by enterokinase, and eluted under non-denaturing conditions.
  • Used as a multifunctional fusion tag
  • Purifies recombinant proteins
  • Maintains natural folding of fusing proteins
  • Can be removed by enterokinase
  • Eluted under non-denaturing conditions
Catalog No. 50-004-26469 Supplier Medchemexpress LLC Supplier No. 25MGHYP0223A
FLAG peptide TFA is the TFA salt form of FLAG peptide. It is a multifunctional fusion tag used for the purification of recombinant proteins. It maintains the natural folding of its fusing proteins and can be removed by enterokinase, and eluted under non-denaturing conditions.
  • Used as a multifunctional fusion tag
  • Purifies recombinant proteins
  • Maintains natural folding of fusing proteins
  • Can be removed by enterokinase
  • Eluted under non-denaturing conditions