Learn More
Medchemexpress LLC HY-P1568A 1mg Medchemexpress, Flagelin 22(TFA) CAS: Purity:>98%
SDP
Catalog No. 501934226
Encompass_Preferred
Medchemexpress, HY-P1568A 1mg Flagelin 22(TFA) CAS: Flagelin 22 TFA (Flagellin 22 TFA), a fragment of bacterial flagellin, is an effective elicitor in both plants and algae. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4226 Supplier Medchemexpress LLC Supplier No. HYP1568A1MG
Medchemexpress, HY-P1568A 1mg Flagelin 22(TFA) CAS: Flagelin 22 TFA (Flagellin 22 TFA), a fragment of bacterial flagellin, is an effective elicitor in both plants and algae. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.