Learn More
G-Biosciences Flagella Stain
SCI_ED
Catalog No. S000760
Change view
Click to view available options
Volume (Metric):
50 mL
Catalog No. Volume (Metric)
S000760 50 mL
1 options
Catalog No. S000760 Supplier G-Biosciences Supplier No. 7862207
Add to Cart
Add to Cart
Flagella Stain is recommended for use in quantitative procedures to stain bacterial flagella and identify their location on the bacterial cell.
Flagella stain is a simple, rapid, qualitative method for detecting bacterial flagella and their shape, length, curvature, arrangement and number on the cell. This is useful in providing taxonomic and identifying information about motile anaerobic bacteria.
Specifications
Volume (Metric) 50 mL
Sterility Sterile
Product Type Gram's Iodine