Learn More
AdipoGen Flagellin,Protein. Source: Salmonella typhimurium
SDP
Supplier:  AdipoGen AG40B0095C100
Encompass_Preferred
Flagellin, AdipoGen Life Sciences. Protein. Source: Salmonella typhimurium strain 14028. Endotoxin content: <0.1EU/?g purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse TLR5 (Toll-like receptor 5). Purity: >90% (SDS-PAGE). Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella. It activates the innate immune system through the receptor Toll-like Receptor 5 (TLR5) and the intracellular protein NLRC4 (NLR family CARD domain-containing protein 4).
Catalog No. 50-464-118