Learn More
Invivogen FLAGELLIN 100MICRONGRAMS
SDP
Supplier:  Invivogen TLRLSTFLA
Encompass
Flagellin TLR5 Ligands, from salmonela thyphimurium, Working Concentration: 0.01 to 1microgram/mL, Quantity: 100microgram
Catalog No. NC9008228