Learn More
AdipoGen Flagellin (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01253010
Encompass_Preferred
Protein. The native flagellin [FliC] from Salmonella enterica Serovar Typhimurium strain ATCC14028 (aa 1-495) is fused at the N-terminus to a His-tag. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Detected by human and mouse TLR5 (Toll-like receptor 5) and mouse NLRC4 (Nod-like receptor [NLR] family caspase activation and recruitment domain [CARD] domain-containing protein 4). Purity: >95% (SDS-PAGE). Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella. It activates the innate immune system through the receptor Toll-like Receptor 5 (TLR5) and the intracellular protein NLRC4 (NLR family CARD domain-containing protein 4).
Catalog No. 50-205-3202