Learn More
AdipoGen Flagellin (TLR5 Mutant) (rec.) (3 x 10 µg)
SDP
Supplier:  AdipoGen AG40B01273010
Encompass_Preferred
Protein. The native flagellin [FliC] from Salmonella enterica Serovar Typhimurium strain ATCC14028 (aa 1-495) contains a substitution of amino acids 90-97 required for TLR5 signaling by a motif from a non-signaling flagellin and is fused at the N-terminus to a His-tag. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Not detected by human and mouse TLR5 (Toll-like receptor 5). Purity: >95% (SDS-PAGE). Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella. It activates the innate immune system through the receptor Toll-like Receptor 5 (TLR5) or the intracellular NLRC4 protein. The Flagellin (TLR5 Mutant) (rec.) cannot be detected by human and mouse TLR5. It has not been tested on NLRC4.
Catalog No. 50-205-3206