Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Flagging Tape

Catalog No. 19104294 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Fluorescent Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-294 Fluorescent Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-294 Supplier Brady™ Supplier No. 58349
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use non-adhesive flagging tape to mark boundaries and hazardous areas

Specifications

Color Fluorescent Red
Width (Metric) 99.06 mm
Packaging Quantity 150 ft./Roll
Height (English) 1.188 in.
Length (English) 150 ft.
Type Barricade Tape
Displayed Text Blank
Material Plastic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.